Learn More
Lambda, Ig Light Chain Mouse anti-Human, FITC, Clone: 1-155-2, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 346600
Description
Anti-Lambda is specific for lambda light chains of human immunoglobulins. Immunoglobulins bearing lambda light chains are present on approximately 40% of normal B lymphocytes and on Igλ+ neoplastic cells. In serum, Anti-Lambda reacts with immunoglobulins bearing lambda light chains as well as free lambda light chains.
Flow Cytometry

Specifications
Lambda, Ig Light Chain | |
Monoclonal | |
12.5μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Human IgA | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
1-155-2 | |
FITC | |
Mouse | |
50 Tests | |
Immunology | |
Human | |
IgG1 λ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.