Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin A/C Mouse anti-Bovine, Canine, Human, Hamster, Mouse, Rat, Sheep, Clone: 131C3, MilliporeSigma™

Catalog No. MAB3538MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3538MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3538MI Supplier MilliporeSigma Supplier No. MAB3538

Mouse Monoclonal Antibody

Anti-Lamin A/C Antibody, localized to aa319-566 rat lamin A, clone 131C3 detects level of Lamin A/C and has been published and validated for use in flow cytometry, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lamin A/C
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 131C3
Conjugate Unconjugated
Formulation Purified culture supernatant. Liquid in PBS containing 0.09% sodium azide.
Gene Accession No. NM_005572.3; NM_170707.2; NM_170708.2
Gene Symbols LMNA, HGPS, EMD2, FPLD, CDCD1, LDP1, LGMD1B, IDC, CMT2B1, FPL, LMNC, PRO1, Lamin-A/C, LMN1, CMD1A, LFP, CDDC
Host Species Mouse
Immunogen Lamin purified from rat liver
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Canine, Hamster, Human, Mouse, Rat, Sheep
Content And Storage −20°C in undiluted aliquots for 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.