Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Lamin B1, Polyclonal, R&D Systems™

Catalog No. AF8525
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8525 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8525 Supplier R&D Systems Supplier No. AF8525
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Specifications

Antigen Lamin B1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunohistochemistry 1:15 dilution, Immunocytochemistry 1:150 dilution, Knockout Validated
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. See Certificate of Analysis for details. with Sodium Azide
Gene Accession No. P20700
Gene Alias ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, LMNB1, MGC111419
Gene Symbols LMNB1
Host Species Rabbit
Immunogen E. coli-derived recombinant human Lamin B1, Met471-Ser582, Accession # P20700
Purification Method Antigen Affinity-purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4001
Test Specificity Detects human Lamin B1 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C, as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 6 months, -20 to -70 °C under sterile conditions after opening.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.