Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin α1/Β1 Rat anti-Mouse, Clone: AL-4, MilliporeSigma™

Catalog No. MAB1905IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1905IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1905IMI Supplier MilliporeSigma Supplier No. MAB1905I
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Laminin α1/β1 Antibody, clone AL-4 is an antibody against Laminin α1/β1 for use in Western Blotting, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin α1/β1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone AL-4
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_032506
Gene Symbols Lamb1, Lamb-1, Lamb1-1, LAMA1, LAMA, PLK-4, PLK4
Host Species Rat
Immunogen Laminin isolated from mouse EHS tumor
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.