Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin-1 A&B chains Rat anti-Mouse, Clone: AL-2, MilliporeSigma™

Catalog No. MAB1904MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1904MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1904MI Supplier MilliporeSigma Supplier No. MAB1904
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect Laminin-1 AandB chains using this Anti-Laminin-1 AandB chains Antibody, cross region, clone AL-2 validated for use in ELISA, immunofluoresence, immunohistochemistry (paraffin), immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin-1 A&B chains
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AL-2
Conjugate Unconjugated
Formulation Purified in 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% NaN3.
Gene Accession No. NM_005559.2
Gene Symbols LAMA1, LAMA
Host Species Rat
Immunogen Laminin isolated from EHS mouse sarcoma.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.