Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin γ1 Mouse anti-Human, Rat, Clone: 2E8, MilliporeSigma™

Catalog No. MAB1920MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1920MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1920MI Supplier MilliporeSigma Supplier No. MAB1920
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Laminin γ1 Antibody, clone 2E8 is validated for use in ELISA, CULT, immunohistochemistry, immunoprecipitation, western blotting for the detection of Laminin γ1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin γ1
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2E8
Conjugate Unconjugated
Formulation Liquid, no preservatives added.
Gene Accession No. NM_002293.3
Gene Symbols LAMC1, LAMB2, MGC87297
Host Species Mouse
Immunogen Purified human laminin
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.