Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Laminin β1 (Laminin B1 Chain), Clone: 4E10, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB1921/DEL
Description
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications
Laminin β1 (Laminin B1 Chain) | |
Unconjugated | |
Purified human laminin | |
RUO | |
Primary | |
Ascites |
Monoclonal | |
Mouse | |
100 μL | |
Cell Structure | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction