Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Laminin-5 Alexa Fluor 647, Clone: D4B5,

Catalog No. MAB19562AF6 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 647
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MAB19562AF6 100 μL Alexa Fluor 647
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB19562AF6 Supplier MilliporeSigma™ Supplier No. MAB19562AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Laminin-5 Clone: D4B5 in Human, Mouse samples, and it is validated for Immunocytochemistry

Laminin subunit gamma-2 (UniProt Q13753; also known as CSF 140kDa subunit, Cell-scattering factor 140kDa subunit, Epiligrin subunit gamma, Kalinin subunit gamma, Ladsin 140kDa subunit, Laminin B2t chain, Lamin-5 subunit gamma, Large adhesive scatter factor 140kDa subunit, Nicein subunit gamma) is encoded by the LAMC2 (also known as B2T, LAMB2T, LAMNB2) gene (Gene ID 3918) in human. Laminins are basement membrane, extracellular matrix glycoproteins composed of alpha, beta and gamma heterotrimeric chains. Different combinations of five alpha chains, three beta chains, and two gamma chains form at least 12 laminin isoforms. Laminin-5 (LN5), which consists of laminin alpha3, beta3 and gamma2 chains, is a highly conserved component of anchoring filaments underlying the hemidesmosome structure of epidermal keratinocytes. Mutations or deletion in the LN5 genes (LAMA3, LAMB3, and LAMC2) is associated with epidermolysis bullosa, a lethal skin blistering disease. LN5 may stabilize the epidermal/dermal junction of skin through interaction with integrins alpha3/beta1, alpha6/beta4 and type VII collagen and has a critical role in the anchorage and locomotion of a wide variety of cell types, including epithelial cells, fibroblasts, neurons and leukocytes. Comparing to fibronectin, collagen, and vitronectin, cells of epithelial origin will adhere to Laminin-5 faster and will spread to a larger extent. Laminin-5 attracted attention when it was detected at the leading edge of invasive cancer tissue in several types of human carcinomas (Niki, T., et al. (2002). Am. J. Pathol. 160(3):112911-41; Lohi, J. (2001). Int. J. Cancer. 94(6):763-767).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin-5
Applications Immunocytochemistry
Classification Monoclonal
Clone D4B5
Conjugate Alexa Fluor 647
Formulation Purified mouse antibody conjugate in PBS with 15mg/mL BSA and 0.1 % sodium azide.
Gene Accession No. Q13753-1
Gene Symbols LAMC2, LAMB2T, LAMNB2
Host Species Mouse
Immunogen Amino acid residues 382-608 of human laminin-5 gamma2 chain (III domain) expressed as a GST fusion protein.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_005553
Test Specificity This antibody recognizes the 150 (precursor form) and 105kDa (mature form) chain proteins of Laminin-5. No reactivity is expected with other laminin types, as the gamma2 chain of laminin is unique to laminin-5.
Target Species Human, Mouse
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.