Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin-5 Mouse anti-Human, AlexaFluor™488, Clone: D4B5, MilliporeSigma™

Catalog No. MAB19562XMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB19562XMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB19562XMI Supplier MilliporeSigma Supplier No. MAB19562X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Laminin-5 Antibody, clone D4B5, Alexa Fluor 488 conjugated is validated for use in flow cytometry, IC for the detection of Laminin-5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin-5
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone D4B5
Conjugate Alexa Fluor 488
Formulation Purified immunoglobulin conjugated to Alexa Fluor™ 488. Liquid in PBS containing 1% BSA, 0.05% Tween™-20 and 0.05% sodium azide.
Gene Accession No. NM_005562.2; NM_018891.2
Gene Symbols LAMC2, EBR2, LAMB2T, MGC141938, MGC138491, LAMNB2, epiligrin, BM600-100kDa, BM600, kalinin-105kDa, nicein-100kDa, EBR2A, B2T, CSF
Host Species Mouse
Immunogen Amino acid residues 382-608 of human laminin gamma2 chain (III domain) expressed as a GST fusion protein.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.