Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin α5 Mouse anti-Human, Clone: 4C7, MilliporeSigma™

Catalog No. MAB1924MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1924MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1924MI Supplier MilliporeSigma Supplier No. MAB1924
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Laminin α5 Antibody, clone 4C7 is an antibody against Laminin α5 for use in ELISA, immunohistochemistry and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin α5
Applications ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 4C7
Conjugate Unconjugated
Formulation UnPurified mouse ascites containing no preservatives.
Gene Accession No. NM_005560.3
Gene Symbols LAMA5, KIAA1907, KIAA0533
Host Species Mouse
Immunogen Purified human laminin
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.