Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB19012MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB19012MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB19012MI Supplier MilliporeSigma Supplier No. AB19012
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Laminin using this Anti-Laminin Antibody validated for use in ELISA, immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid at 1mg/mL in 0.125M Sodium Borate, 0.075M Sodium Chloride, 0.005M EDTA, pH 8.0 with 0.01% Sodium Azide.
Gene Symbols LAMA1; LAMA
Host Species Rabbit
Immunogen Full length human laminin. Immunization occurred after single-band purity was assessed by SDS-PAGE.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005559.2
Target Species Human
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.