Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin B2 Rat anti-Human, Mouse, Clone: A5, MilliporeSigma™

Catalog No. 05206MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-206-MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-206-MI Supplier MilliporeSigma Supplier No. 05206
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Laminin B2 Antibody, clone A5 is an antibody against Laminin B2 for use in immunoprecipitation, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin B2
Applications Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone A5
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.0
Gene Accession No. NM_002292
Gene Symbols LAMB2, S-laminin, LAMS
Host Species Rat
Immunogen Murine EHS laminin
Purification Method Protein G chromatography
Quantity 50 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.