Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamp-1 Mouse anti-Human, Unlabeled, Clone: 25, BD

Catalog No. BDB611043 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611043 150 μg
BDB611042 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611043 Supplier BD Biosciences Supplier No. 611043
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lysosomes are digestive compartments in mammalian cells that are responsible for the degradation of numerous substances including foreign materials. The lysosomal membrane is of interest due to its role in maintaining the acidic intralysosomal sac, its resistance to degradation, and its ability to fuse with other membranous organelles. Lamp-1 and lamp-2 (lysosomal-associated membrane proteins -1 and -2) were identified as lysosomal membrane proteins that act as carriers for poly-N-glycans, primarily N-acetyllactosamines. They consist of two heavily glycosylated amino terminal domains, a single trans-membrane domain, and a short cytoplasmic tail. A tyrosine located at the fourth residue from the end of the cytoplasmic tail, a large hydrophobic amino acid as the last residue, and a glycine residue amino terminal to the tyrosine residue targets these proteins from the TGN to the lysosome. The role of the Lamp molecules may be a carrier for poly-N-acetyllactosamines and/or to protect lysosomal membranes from the soluble hydrolases.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lamp-1
Applications Western Blot
Classification Monoclonal
Clone 25
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Lamp-1 aa. 25-224
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.