Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP-2 Rat anti-Mouse, Clone: ABL-93, MilliporeSigma™

Catalog No. MABC40MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC40MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC40MI Supplier MilliporeSigma Supplier No. MABC40
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-LAMP-2 Antibody, clone ABL-93 is an antibody against LAMP-2 for use in western blot, immunoprecipitation and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAMP-2
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone ABL-93
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001017959
Gene Symbols CD107b, LAMPB, LGP110, LAMP-2
Host Species Rat
Immunogen Purified glycoproteins from NIH/3T3 cells, corresponding to mouse LAMP-2.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.