Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAP Mouse anti-Human, PerCP-Cy5.5, Clone: TW4-2F8, BD

Catalog No. BDB562544 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562544 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562544 Supplier BD Biosciences Supplier No. 562544
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TW4-2F8 monoclonal antibody specifically binds to Latency-Associated Peptide (LAP), a component of the dimeric Transforming Growth Factor-beta 1 (TGF-β1) propeptide encoded by TGFB1. Prior to secretion, the dimeric LAP-TGF-β1 propeptide is cleaved resulting in a biologically inactive form of dimeric TGF-β1 that is noncovalently associated with dimeric LAP (latent TGF-β1). This complex may be expressed on the surface of TGF-β1TGF-β1. TGF-β1 is a potent multifunctional cytokine that regulates numerous processes including development, hematopoiesis, tissue remodeling, wound repair, and immunity as well as cancer and autoimmune diseases. Clone TW4-2F8 is routinely quality tested through intracellular staining of LAP in P3UI-TGFβ1 transfected cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen LAP
Applications Flow Cytometry
Classification Monoclonal
Clone TW4-2F8
Conjugate PerCP-Cyanine5.5
Description Latency-associated peptide; TGFB1; TGFbeta; Transforming growth factor beta
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human TGF-β1
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.