Learn More
LAP Mouse anti-Human, PE-CF594, Clone: TW4-2F8, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 562490
Description
The TW4-2F8 monoclonal antibody specifically binds to Latency-Associated Peptide (LAP), a component of the dimeric Transforming Growth Factor-beta 1 ( TGF-β1 ) propeptide encoded by TGFB1 . Prior to secretion, the dimeric LAP- TGF-β1 propeptide is cleaved resulting in a biologically inactive form of dimeric TGF-β1 that is noncovalently associated with dimeric LAP (latent TGF-β1 ). This complex may be expressed on the surface of TGF-β1 TGF-β1 . TGF-β1 is a potent multifunctional cytokine that regulates numerous processes including development, hematopoiesis, tissue remodeling, wound repair, and immunity as well as cancer and autoimmune diseases. Clone TW4-2F8 is routinely quality tested through intracellular staining of LAP in P3UI-TGFβ1 transfected cells.
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).

Specifications
LAP | |
Monoclonal | |
PE-CF594 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human TGF-β1 | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
TW4-2F8 | |
Latency-associated peptide; TGFB1; TGFbeta; Transforming growth factor beta | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.