Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAP2 Mouse, Unlabeled, Clone: 27, BD

Catalog No. BDB611000 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611000 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611000 Supplier BD Biosciences Supplier No. 611000
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A specialized extension of the ER, the nuclear envelope (NE) forms the nuclear compartment boundary in eukaryotic cells. It contains numerous pore complexes and the nucleoplasmic side is linked to nuclear lamina. The nuclear lamina composes the structural framework for the NE and serves as a chromatin anchor site, thus, playing a major role in interphase nuclear organization. Many proteins are associated with lamina, particularly the LAPs (Lamina-Associated Polypeptides). LAP2 (also known as LAP2 β; ) is a hydrophilic protein with a single transmembrane segment near the C-terminus. Thus, it has been defined as a type II integral membrane protein with the majority of its structure exposed to the nucleoplasm. LAP2 binding to lamins contributes to the attachment of the nuclear lamina to the inner nuclear membrane. LAP2 also binds to chromatin, implying its role in chromosomal organization during mitosis. Mitotic phosphorylation of LAP2 regulates its binding to lamins and chromosomes during the disassembly and reassembly of mitosis. Thus, LAP2 is a nuclear protein that plays a role in the organization of the NE during cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAP2
Applications Western Blot
Classification Monoclonal
Clone 27
Concentration 250μg/mL
Conjugate Unconjugated
Description LAP2β; Lamina-Associated Polypeptides
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat LAP2 aa. 34-156
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.