Learn More
LAP2 Rabbit anti-Human, Canine, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 061002
Description
Anti-LAP2 Antibody is an antibody against LAP2 for use in western blot, IC and immunofluoresence.
Specifications
LAP2 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine, (pH 7.4), 150mM NaCl, with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Canine, Human, Mouse, Rat |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
CMD1T ; LAP2 ; LEMD4; MGC61508 ; PRO0868; TP; thymopoietin | |
KLH-conjugated synthetic linear peptide corresponding to LAP2. | |
100 μL | |
NM_003276 | |
2-8°C for 12 months |