Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LASP-1 Mouse anti-Human, Clone: 8C6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB8991
Description
Anti-LASP-1 Antibody, clone 8C6 is an antibody against LASP-1 for use in ELISA, immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry.
Specifications
LASP-1 | |
Monoclonal | |
Unconjugated | |
NM_006148.1 | |
Mouse | |
100 μL | |
Human | |
Ascites |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
8C6 | |
Ascites fluid. Liquid containing no preservatives. | |
LASP1, LASP-1, MLN50, Lasp-1 | |
Synthetic peptide corresponding to an internal sequence of human Lasp-1. | |
Primary | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction