Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAG1 homolog, ceramide synthase 1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89018859 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-859 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-859 Supplier Abnova Corporation Supplier No. H00010715A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant LASS1.

This gene encodes a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site that is cleaved to produce a mature protein containing seven conserved cysteine residues. Members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in yeast suggest that the encoded protein is involved in aging. This protein is transcribed from a monocistronic mRNA as well as a bicistronic mRNA, which also encodes growth differentiation factor 1. [provided by RefSeq

Sequence: YIVAFAAKVLTGQVHELKDLREYDTAEAQSLKPSKAEKPLRNGLVKDKRF

Specifications

Antigen LAG1 homolog, ceramide synthase 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant LASS1.
Formulation 50% glycerol
Gene LASS1
Gene Accession No. NM_021267
Gene Alias CerS1/LAG1/MGC90349/UOG1
Gene Symbols LASS1
Host Species Mouse
Immunogen LASS1 (NP_067090, 301 a.a. ∼ 350 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10715
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.