Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAT Mouse anti-Human, Clone: 2E9, MilliporeSigma™

Catalog No. 05561MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-561-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-561-MI Supplier MilliporeSigma Supplier No. 05561
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect LAT using this Anti-LAT Antibody, clone 2E9 validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAT
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2E9
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in Protein A Purified immunoglobulin in 0.1M Tris-Glycine, 0.15M NaCl and 0.05% Sodium Azide.
Gene Accession No. NM_014387
Gene Symbols LAT, pp36, LAT1, p36-38
Host Species Mouse
Immunogen GST fusion protein corresponding to residues 31-233 of human LAT
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.