Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAT Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 06807MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
400 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-807-MI 400 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-807-MI Supplier MilliporeSigma Supplier No. 06807
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-LAT Antibody detects level of LAT and has been published and validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAT
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, and 0.05% sodium azide.
Gene Symbols LAT; LAT1; pp36; p36-38
Host Species Rabbit
Immunogen GST-tagged fusion protein corresponding to residues 31-233 of human LAT
Purification Method Protein A purfied
Quantity 400 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_014387
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.