Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-LBPA Clone: 6C4,

Catalog No. MABT837MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABT837MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT837MI Supplier MilliporeSigma™ Supplier No. MABT837
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects LBPA Clone: 6C4 in All samples, and it is validated for Dot Blot, Electron Microscopy, ELISA, Immunocytochemistry

Lyso bis-phosphatidic acid (LBPA, Lysobisphosphatidic acid, L(bis)PA) is a lipid that is exclusively located in late endosomes and is used as a late endosome marker. One of the characteristic features of late endosomes is a complex system of internal membranes within the lumen that contains large amounts of the unique, poorly degradable LBPA, and thus forms a specialized membrane domain within endosomes. Late endosomes function not only as a major protein-sorting compartment, but also an obligatory station for LDL and other endocytosed ligands destined for degradation. Research shows that the LBPA-rich membranes within late endosomes regulate cholesterol transport, presumably by acting as a collection and distribution device. The genetic disease Niemann-Pick type C (NPC) is characterized by both lysosomal and endosomal storage disorders. In skin fibroblasts from NPC patients, cholesterol accumulation is seen within vesicles containing late endosome markers LBPA and Rab7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LBPA
Applications Dot Blot, Immunoassay, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 6C4
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Endosomal membranes from baby hamster kidney fibroblasts (BHK) corresponding to Hamster LBPA.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone 6C4 specifically detects LBPA, but not PC, PE, SM, PS, CL, PI, Sul, Chol, Cer, or CE purified from baby hamster kidney (BHK) cell lipid extract (Kobayahsi, T., et al. (1998). Nature. 392(6672):193-197).
Target Species All species
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.