Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lbx1 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB15292MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15292MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15292MI Supplier MilliporeSigma Supplier No. AB15292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Lbx1 Antibody is validated for use in western blotting for the detection of Lbx1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lbx1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in PBS with 0.1% sodium azide.
Gene Symbols LBX1; homeobox; LBX1H; HPX6; HPX-6
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Immunoaffinity purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_006562.4
Target Species Mouse
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.