Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3B Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABC432MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC432MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC432MI Supplier MilliporeSigma Supplier No. ABC432
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect MAP1 using this rabbit polyclonal antibody, Anti-LC3B Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LC3B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Symbols MAP1LC3B; MAP1ALC3
Host Species Rabbit
Immunogen Linear peptide corresponding to human LC3B.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_073729
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.