Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lck Mouse anti-Human, Mouse, Rat, Clone: 3A5, MilliporeSigma™

Catalog No. 05435MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-435-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-435-MI Supplier MilliporeSigma Supplier No. 05435
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Lck Antibody, clone 3A5 is validated for use in immunoprecipitation, western blotting for the detection of Lck.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lck
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3A5
Conjugate Unconjugated
Formulation Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide as a preservative.
Gene Accession No. NM_005356
Gene Symbols LCK, pp58lck, LSK, p56-LCK, p56lck, YT16
Host Species Mouse
Immunogen GST-fusion to residues 1-225 of murine Lck
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.