Learn More
lymphocyte-specific protein tyrosine kinase, Mouse, Clone: 8B10, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene is a member of the Src family of protein tyrosine kinases (PTKs). The encoded protein is a key signaling molecule in the selection and maturation of developing T-cells. It contains N-terminal sites for myristylation and palmitylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. The protein localizes to the plasma membrane and pericentrosomal vesicles, and binds to cell surface receptors, including CD4 and CD8, and other signaling molecules. Multiple alternatively spliced variants, encoding the same protein, have been described. [provided by RefSeq
Specifications
Specifications
| Antigen | lymphocyte-specific protein tyrosine kinase |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 8B10 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant LCK. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | LCK |
| Gene Accession No. | BC013200 |
| Gene Alias | YT16/p56lck/pp58lck |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.