Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIM domain binding 1, Mouse, Clone: 4G8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant LDB1.
Supplier: Abnova Corporation H00008861M03
Description
Sequence: MLDRDVGPTPMYPPTYLEPGIGRHTPYGNQTDYRIFELNKRLQNWTEECDNLWWDAFTTEFFEDDAMLTITFCLEDGPKRYTIGRTLIPRYFRSIFEGGATELYYVLKHPSpecifications
LIM domain binding 1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_003893 | |
LDB1 | |
LDB1 (NP_003884, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Transcription Regulation | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
4G8 | |
Mouse monoclonal antibody raised against a partial recombinant LDB1. | |
LDB1 | |
CLIM2/NLI | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
8861 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction