Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEF-1 Mouse anti-Human, Clone: 1C3.1D10, MilliporeSigma™

Catalog No. MAB3750MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3750MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3750MI Supplier MilliporeSigma Supplier No. MAB3750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-LEF-1 Antibody, all isoforms, clone 1C3.1D10 is an antibody against LEF-1 for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEF-1
Applications Western Blot
Classification Monoclonal
Clone 1C3.1D10
Conjugate Unconjugated
Formulation Purified immunoglobulin. Provided as solution in phosphate buffered saline with 0.08% sodium azide.
Gene Accession No. NM_016269.2
Gene Symbols LEF1, LEF-1, TCF1-alpha, TCF1ALPHA, DKFZp586H0919
Host Species Mouse
Immunogen Recombinant full length LEF-1 protein.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.