Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEF-1 Mouse anti-Human, Clone: 3A12, MilliporeSigma™

Catalog No. MAB3749MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3749MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3749MI Supplier MilliporeSigma Supplier No. MAB3749
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-LEF-1 Antibody, transactivation domain, clone 3A12 is validated for use in western blotting for the detection of LEF-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LEF-1
Applications Western Blot
Classification Monoclonal
Clone 3A12
Conjugate Unconjugated
Formulation Purified immunoglobulin. Provided as 0.2μm sterile filtered solution in phosphate buffered saline with 0.08% sodiumazide.
Gene Accession No. NM_016269.2
Gene Symbols LEF1, LEF-1, TCF1-alpha, TCF1ALPHA, DKFZp586H0919
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from mice immunized with synthetic peptide corresponding to amino acids 236-242 of human LEF-1 and mouse myeloma cells.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.