Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

lymphoid enhancer-binding factor 1, Mouse, Clone: 5A3, Abnova™

Catalog No. 89001278 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-278 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-278 Supplier Abnova Corporation Supplier No. H00051176M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant LEF1.

LEF1 is a nuclear protein that is expressed in pre-B and T cells. It binds to a functionally important site in the T-cell receptor-alpha (TCRA; MIM 186880) enhancer and confers maximal enhancer activity. LEF1 belongs to a family of regulatory proteins that share homology with high mobility group protein-1 (HMG1; MIM 163905) (Waterman et al., 1991 [PubMed 2010090]; van Genderen et al., 1994 [PubMed 7958926]).[supplied by OMIM

Sequence: DPELCATDEMIPFKDEGDPQKEKIFAEISHPEEEGDLADIKSSLVNESEIIPASNGHEVARQAQTSQEPYHDKAREHPDDGKHPDGGLYNKGPSYSSYSGYIMMPNMNND

Specifications

Antigen lymphoid enhancer-binding factor 1
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5A3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant LEF1.
Formulation PBS with no preservative; pH 7.4
Gene LEF1
Gene Accession No. NM_016269
Gene Alias DKFZp586H0919/TCF1ALPHA
Gene Symbols LEF1
Host Species Mouse
Immunogen LEF1 (NP_057353, 14 a.a. ∼ 123 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 51176
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.