Learn More
left-right determination factor 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human LEFTY2 protein.
Supplier: Abnova Corporation H00007044D01P
Description
This gene encodes a member of the TGF-beta family of proteins. The encoded protein is secreted and plays a role in left-right asymmetry determination of organ systems during development. The protein may also play a role in endometrial bleeding. Mutations in this gene have been associated with left-right axis malformations, particularly in the heart and lungs. Some types of infertility have been associated with dysregulated expression of this gene in the endometrium. Alternative processing of this protein can yield three different products. This gene is closely linked to both a related family member and a related pseudogene. [provided by RefSeq
Sequence: MWPLWLCWALWVLPLAGPGAALTEEQLLGSLLRQLQLSEVPVLDRADMEKLVIPAHVRAQYVVLLRRSHGDRSRGKRFSQSFREVAGRFLASEASTHLLVFGMEQRLPPNSELVQAVLRLFQEPVPKAALHRHGRLSPRSAQARVTVEWLRVRDDGSNRTSLIDSRLVSVHESGWKAFDVTEAVNFWQQLSRPRQPLLLQVSVQREHLGPLASGAHKLVRFASQGAPAGLGEPQLELHTLDLRDYGAQGDCDPEAPMTEGTRCCRQEMYIDLQGMKWAKNWVLEPPGFLAYECVGTCQQPPEALAFNWPFLGPRQCIASETASLPMIVSIKEGGRTRPQVVSLPNMRVQKCSCASDGALVPRRLQPSpecifications
left-right determination factor 2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human LEFTY2 protein. | |
LEFTY2 | |
EBAF/LEFTA/LEFTYA/MGC46222/TGFB4 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_003240 | |
LEFTY2 | |
LEFTY2 (NP_003231.2, 1 a.a. ∼ 366 a.a) full-length human protein. | |
100 μg | |
Cytokines | |
7044 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.