Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LEO1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079777
Description
LEO1 Polyclonal specifically detects LEO1 in Human, Mouse samples. It is validated for Western Blot.Specifications
LEO1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
Leo1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae), RDLReplicative senescence down-regulated leo1-like protein, RNA polymerase-associated protein LEO1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:1000-1:5000, Immunoprecipitation | |
Q8WVC0 | |
LEO1 | |
The immunogen recognized by this antibody maps to a region between residues 350 and 400 of human senescence down regulated leo1-like using the numbering given in TrEMBL entry Q8WVC0 (GeneID 123169). | |
100 μL | |
Transcription Factors and Regulators | |
123169 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction