Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEPRE1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017148 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-148 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-148 Supplier Abnova Corporation Supplier No. H00064175B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human LEPRE1 protein.

This gene encodes an enzyme that is a member of the collagen prolyl hydroxylase family. These enzymes are localized to the endoplasmic reticulum and their activity is required for proper collagen synthesis and assembly. Mutations in this gene are associated with osteogenesis imperfecta type VIII. Two alternatively spliced transcript variants encoding different isoforms have been described. Other variants may exist, but their biological validity has not been determined. [provided by RefSeq

Sequence: MLGEEHTRSIGPRESAKEYRQRSLLEKELLFFAYDVFGIPFVDPDSWTPEEVIPKRLQEKQKSERETAVRISQEIGNLMKEIETLVEEKTKESLDVSRLTREGGPLLYEGISLTMNSKLLNGSQRVVMDGVISDHECQELQRLTNVAATSGDGYRGQTSPHTPNEKFYGVTVFKALKLGQEGKVPLQSAHLYYNVTEKVRRIMESYFRLDTPLYFSYSHLVCRTAIEEVQAERKDDSHPVHVDNCILNAETLVCVKEPPAYTFRDYSAILYLNGDFDGGNFYFTELDAKTVTAEVQPQCGRAVGFSSGTENPHGVKAVTRGQRCAIALWFTLDPRHSERDRVQADDLVKMLFSPEEMDLSQEQPLDAQQGPPEPAQESLSGSESKPKDEL

Specifications

Antigen LEPRE1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human LEPRE1 protein.
Formulation PBS with no preservative; pH 7.4
Gene LEPRE1
Gene Accession No. BC015309.1
Gene Alias GROS1/MGC117314/P3H1
Gene Symbols LEPRE1
Host Species Mouse
Immunogen LEPRE1 (AAH15309, 1 a.a. ∼ 390 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64175
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.