Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leptin Receptor (CD295) Mouse anti-Human, Alexa Fluor 647, Clone: 52263, BD

Catalog No. BDB564376 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564376 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564376 Supplier BD Biosciences Supplier No. 564376
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 52263 monoclonal antibody specifically binds to the Leptin Receptor (Leptin R/LEPR) which is also known as CD295. CD295 is a type I transmembrane glycoprotein that belongs to the type I Cytokine Receptor family within the Immunoglobulin gene superfamily. Multiple isoforms of CD295 have been detected including a long form with a large cytoplasmic domain capable of signal transduction. Several receptor isoforms with short cytoplasmic domains that lack the capacity to transduce signals have also been reported. CD295 is widely expressed by a variety of cells within different tissues. CD295 binds to leptin with high affinity and plays important roles in lipid metabolism, fertility, angiogenesis, bone formation and immunity.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Leptin Receptor (CD295)
Applications Flow Cytometry
Classification Monoclonal
Clone 52263
Conjugate Alexa Fluor 647
Description LEPR; Leptin receptor; LEP-R; OB receptor; OBR; HuB219
Host Species Mouse
Immunogen Human Leptin Receptor Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.