Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leukemia-associated RhoGEF/LARG Mouse anti-Human, Clone: 13D2.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT124
Description
Anti-Leukemia-associated RhoGEF/LARG Antibody, clone 13D2.1 is a highly specific mouse monoclonal antibody, that targets Leukemia-associated RhoGEF and has been tested in western blotting and immunocytochemistry.
Specifications
Leukemia-associated RhoGEF/LARG | |
Monoclonal | |
Unconjugated | |
NP_056128 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG2b κ |
Immunocytochemistry, Western Blot | |
13D2.1 | |
Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
ARHGEF12, KIAA0382, LARG | |
GST-tagged recombinant protein corresponding to the RGSL domain of human Leukemia-associated RhoGEF/LARG. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction