Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Leukemia Inhibitory Factor Mouse anti-Mouse, Clone: 2H2.2, MilliporeSigma™

Catalog No. MAB4307MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4307MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4307MI Supplier MilliporeSigma Supplier No. MAB4307
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Mouse Leukemia Inhibitory Factor Antibody, clone 2H2.2 is an antibody against Mouse Leukemia Inhibitory Factor for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Leukemia Inhibitory Factor
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2H2.2
Conjugate Unconjugated
Formulation Purified Immunoglobulin. Liquid in 0.02M PBS, 0.25M NaCl ph 7.6 with 0.1% Sodium Azide.
Gene Symbols LIF, HILDA, MLPLI, CDF
Host Species Mouse
Immunogen Recombinant Murine LIF.
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.