Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Leukemia Inhibitory Factor Mouse anti-Human, Mouse, Clone: 4F7.2, MilliporeSigma™

Catalog No. MAB4306MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4306MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4306MI Supplier MilliporeSigma Supplier No. MAB4306
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Human Leukemia Inhibitory Factor using this Anti-Human Leukemia Inhibitory Factor Antibody, clone 4F7.2 validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Leukemia Inhibitory Factor
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4F7.2
Conjugate Unconjugated
Gene Accession No. NM_002309.2
Gene Symbols LIF, Emfilermin, CDF, MLPLI, HILDA, D-FACTOR
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.