Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leukemia Inhibitory Factor Mouse anti-Human, PE, Clone: 1F10, BD

Catalog No. BDB558571 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558571 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558571 Supplier BD Biosciences Supplier No. 558571
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1F10 antibody specifically binds to human Leukemia Inhibitory Factor (LIF) also known as Human Interleukin for DA cells (HILDA), Melanoma-derived Lipoprotein Lipase Inhibitor (MLPLI) and Hepatocyte Stimulating Factor III (HSF III). LIF is produced by multiple sources including activated T cells and macrophages, myelomonocytic lineages, fibroblasts, liver, heart and melanoma cells. LIF regulates the differentiation of embryonic stem cells, neural cells, osteoblasts, adipocytes, hepatocytes and kidney epithelial cells. Other activities include terminal differentiation in leukemic cells and the stimulation of acute-phase protein synthesis in hepatocytes. Many of its biological functions parallel those of Interleukin-6, Oncostatin M, Ciliary Neurotrophic Factor, Interleukin-11 and Cardiotrophin-1. In vivo LIF is important in regulating the inflammatory response by tuning the balance of four systems in the body, namely the immune, the haematopoietic, the nervous and the endocrine systems. The immunogen used to generate the 1F10 hybridoma was recombinant vaccinia virus encoding the human LIF cytokine. The LIF cytokine had been engineered to get expressed on the membrane of the infected cell. The attachment to the membrane was obtained with the glycosylphoshosphatidyl anchor targeting DNA sequence from the DAF molecule (CD55). This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Leukemia Inhibitory Factor
Applications Flow Cytometry
Classification Monoclonal
Clone 1F10
Conjugate PE
Description MLPLI; HILDA; HSF III
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant vaccinia virus
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.