Learn More
anti-Lewis A (Blood Group Antigen) Ab-1, Clone: LWA01; same as 7LE, Epredia™ LabVision
Shop All Epredia ProductsDescription
Lewis A is a blood group antigen consisting of Gal Β1-3(Fuc α1-4) GlcNAc. Lewis A is found to be present primarily on epithelial cells such as colon and kidneys.2-4 In the tumors and dedifferentiated tissues, decrease of Lea antigen was observed.
Host Species: Mouse
Clone: LWA01; same as 7LE
Isotype: IgGa/κ
Cross Reactivity: Human. Does not react with rat. Others not-tested.
Epitope: Gal Β1-3(Fuc α1-4) GlcNAc
Positive Control: Colon
Cellular Localization: Cytoplasmic
Immunohistochemistry (Formalin/Paraffin)
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
| Antigen | Lewis A (Blood Group Antigen) Ab-1 |
| Classification | Monoclonal |
| Concentration | — |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 7 mL |
| Primary or Secondary | Primary |
| Target Species | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.