Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LeX Glycan Mouse anti-Rat, Clone: Forse-1, MilliporeSigma™

Catalog No. MABD125MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD125MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD125MI Supplier MilliporeSigma Supplier No. MABD125

Mouse Monoclonal Antibody

Anti-LeX Glycan Antibody, clone Forse-1 is a highly specific mouse monoclonal antibody, that targets Lewis X and has been tested in western blotting, immunohistochemistry and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LeX Glycan
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Forse-1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ supernatant in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. N/A
Host Species Mouse
Immunogen Whole NT2/D1 cells (human embryonal carcinoma cell line) differentiated with retinoic acid
Purification Method Purified by ion-exchange chromatography
Quantity 100 μL
Primary or Secondary Primary
Target Species Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.