Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

lectin, galactoside-binding, soluble, 3 binding protein, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016835 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-835 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-835 Supplier Abnova Corporation Supplier No. H00003959B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human LGALS3BP protein.

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP has been found elevated in the serum of patients with cancer and in those infected by the human immunodeficiency virus (HIV). It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Using fluorescence in situ hybridization the full length 90K cDNA has been localized to chromosome 17q25. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1. [provided by RefSeq

Sequence: MTPPRLFWVWLLVAGTQGVNDGDMRLADGGATNQGRVEIFYRGQWGTVCDNLWDLTDASVVCRALGFENATQALGRAAFGQGSGPIMLDEVQCTGTEASLADCKSLGWLKSNCRHERDAGVVCTNETRSTHTLDLSRELSEALGQIFDSQRGCDLSISVNVQGEDALGFCGHTVILTANLEAQALWKEPGSNVTMSVDAECVPMVRDLLRYFYSRRIDITLSSVKCFHKLASAYGARQLQGYCASLFAILLPQDPSFQMPLDLYAYAVATGDALLEKLCLQFLAWNFEALTQAEAWPSVPTDLLQLLLPRSDLAVPSELALLKAVDTWSWGERASHEEVEGLVEKIRFPMMLPEELFELQFNLSLYWSHEALFQKKTLQALEFHTVPFQLLARYKGLNLTEDTYKPRIYTSPTWSAFVTDSSWSARKSQLVYQSRRGPLVKYSSDYFQAPSDYRYYPYQSFQTPQHPSFLFQDKRVSWSLVYLPTIQSCWNYGFSCSSDELPVLGLTKSGGSDRTIAYENKALMLCEGLFVADVTDFEGWKAAIPSALDTNSSKSTSSFPCPAGHFNGFRTVIRPFYLTNSSGVD

Specifications

Antigen lectin, galactoside-binding, soluble, 3 binding protein
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human LGALS3BP protein.
Formulation PBS with no preservative; pH 7.4
Gene LGALS3BP
Gene Accession No. NM_005567.2
Gene Alias 90K/BTBD17B/MAC-2-BP
Gene Symbols LGALS3BP
Host Species Mouse
Immunogen LGALS3BP (NP_005558.1, 1 a.a. ∼ 585 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Adhesion
Primary or Secondary Primary
Gene ID (Entrez) 3959
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.