Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-LGALS9, Polyclonal,

Catalog No. ABC262MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC262MI 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC262MI Supplier MilliporeSigma™ Supplier No. ABC262

Specifications

Antigen LGALS9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with 0.1% sodium azide and 50% glycerol.
Gene Symbols LGALS9
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to human LGALS9.
Purification Method Affinity Purified
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 3965
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.