Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGI1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002814 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-814 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-814 Supplier Abnova Corporation Supplier No. H00009211B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human LGI1 protein.

This gene is rearranged as a result of translocations in glioblastoma cell lines. The protein contains a hydrophobic segment representing a putative transmembrane domain with the amino terminus located outside the cell. It also contains leucine-rich repeats with conserved cysteine-rich flanking sequences. This gene is predominantly expressed in neural tissues and its expression is reduced in low grade brain tumors and significantly reduced or absent in malignant gliomas. Mutations in this gene result in autosomal dominant lateral temporal epilepsy. [provided by RefSeq

Sequence: MESERSKRMGNACIPLKRIAYFLCLLSALLLTEGKKPAKPKCPAVCTCTKDNALCENARSIPRTVPPDVISLSFVRSGFTEISEGSFLFTPSLQLLLFTSNSFDVISDDAFIGLPHLEYLFIENNNIKSISRHTFRGLKSLIHLSLANNNLQTLPKDIFKGLDSLTNVDLRGNSFNCDCKLKWLVEWLGHTNATVEDIYCEGPPEYKKRKINSLSSKDFDCIITEFAKSQDLPYQSLSIDTFSYLNDEYVVIAQPFTGKCIFLEWDHVEKTFRNYDNITGTSTVVCKPIVIETQLYVIVAQLFGGSHIYKRDSFANKFIKIQDIEILKIRKPNDIETFKIENNWYFVVADSSKAGFTTIYKWNGNGFYSHQSLHAWYRDTDVEYLEIVRTPQTLRTPHLILSSSSQRPVIYQWNKATQLFTNQTDIPNMEDVYAVKHFSVKGDVYICLTRFIGDSKVMKWGGSSFQDIQRMPSRGSMVFQPLQINNYQYAILGSDYSFTQVYNWDAEKAKFVKFQELNVQAPRSFTHVSINKRNFLFASSFKGNTQIYKHVIVDLSA

Specifications

Antigen LGI1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human LGI1 protein.
Formulation PBS with no preservative; pH 7.4
Gene LGI1
Gene Accession No. NM_005097.1
Gene Alias EPITEMPIN/EPT/ETL1/IB1099
Gene Symbols LGI1
Host Species Mouse
Immunogen LGI1 (NP_005088.1, 1 a.a. ∼ 557 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9211
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.