Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Lgr5 (Central LRR); Clone: 4D11F8; BD

Catalog No. BDB562732 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562732 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562732 Supplier BD Biosciences Supplier No. 562732
Add to Cart
Add to Cart

Host Species: Rat
Clone: 4D11F8
Isotype: IgG2b, κ
Cross Reactivity Species: Hu, Ms
Immunogen: Human LGR5 DNA

Flow Cytometry, Bioimaging, Immunofluorescence, Western Blotting, Immunohistochemistry (Formalin-Antigen Retrieval Required), Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Human Lgr5 (Central LRR)
Classification Monoclonal
Clone 4D11F8
Concentration 0.5mg/mL
Conjugate Unconjugated
Host Species Rat
Immunogen Human LGR5 DNA
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.