Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lgr5 (CRL Region) Rat anti-Human, Unlabeled, Clone: 1D9A5, BD

Catalog No. BDB562733 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562733 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562733 Supplier BD Biosciences Supplier No. 562733
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Lgr5 (leucine-rich-repeat-containing G-protein-coupled receptor 5) is a seven transmembrane-domain receptor that is a target gene for Wnt and marks stem cells in the small intestine and colon, stomach, and hair follicle. Lgr5 was initially identified as a potential stem cell marker due to restricted expression of Lgr5 in the intestinal crypt and labeling of rapidly cycling cells of the colon and intestine. Using both lineage tracing and organiod culture experiments, Lgr5 positive cells are capable of generating all types of the small intestine epithelium hence indicating that Lgr5 marks stem cells of the small intestine and colon. R-spondin growth factors, which are secreted agonists of the Wnt pathway, bind Lgr5. The binding of R-spondins to Lgr5 leads to recruitment of the Frizzled/LRP Wnt receptor complex, which binds to Wnt ligands and leads to downstream Wnt signaling. Lgr5 is up-regulated in colon and ovarian cancers and has been implicated in promotion of tumor growth and metastasis.
The 1D9A5 monoclonal antibody recognizes an epitope in the Cysteine-rich linker (CRL) region of Human Lgr5.

Host Species: Mouse
Clone: 1D9A5
Isotype: IgG2b, λ
Species Reactivity: Hu
Immunogen: Human LGR5 DNA

Bioimaging, Flow Cytometry, Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Lgr5 (CRL Region)
Applications Western Blot
Classification Monoclonal
Clone 1D9A5
Concentration 0.5mg/mL
Conjugate Unconjugated
Description GPR49, GPR67, HG38
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Human LGR5 DNA
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.