Learn More
Description
Host Species: Rat
Clone: 8F2
Isotype: IgG2b, κ
Cross Reactivity Species: Hu, Ms
Immunogen: Human LGR5 DNA
Flow Cytometry, Bioimaging, Immunofluorescence, Western Blotting, Immunohistochemistry (Formalin-Antigen Retrieval Required), Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | Human Lgr5 (N-Terminal) |
| Classification | Monoclonal |
| Clone | 8F2 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Host Species | Rat |
| Immunogen | Human LGR5 DNA |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.