Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LHFPL3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525625
Description
LHFPL3 is a member of the lipoma HMGIC fusion partner(LHFP) gene family, which is a subset of the superfamily oftetraspan transmembrane protein encoding genes. Mutations in oneLHFP-like gene result in deafness in humans and mice, and a secondLHFP-like gene is fused to a high-mobility group gene in atranslocation-associated lipoma. A partial gene fragment namedLHFPL4 corresponds to a portion of the first exon of this gene.
Specifications
LHFPL3 | |
Polyclonal | |
LHFPL3 | |
LHFPL4, LHFP-like protein 3, lipoma HMGIC fusion partner-like 3 protein, lipoma HMGIC fusion partner-like 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
269629, 375612 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q86UP9, Q9CTN8 | |
LHFPL3 | |
KLH conjugated synthetic peptide between 184-213 amino acids from the C-terminal region of human LHPL3 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction