Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-LHX2, Polyclonal

Catalog No. ABE1402MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1402MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1402MI Supplier MilliporeSigma™ Supplier No. ABE1402
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects LHX2 Clone: in Human, Mouse samples, and it is validated for ChIP-seq, Chromatin Immunoprecipitation, Immunofluorescence, Western Blotting

LIM/homeobox protein Lhx2 (UniProt Q9Z0S2; also known as Homeobox protein LH-2, LIM homeo box protein 2, Lim2) is encoded by the Lhx2 gene (Gene ID 16870) in murine species. Lhx2 is a transcription factor that plays an important role in controlling the behavior of progenitor cells involved in the development of the skin hair cycling, eye, forebrain, limb, hematopoietic, olfactory, and pituitary systems. Lhx2 is shown to regulate a significant number of hair follicle stem cell (HF-SC) signature genes, as well as cytoskeletal and adhesion molecules within the niche where HF-SCs reside as a single layer of polarized, quiescent epithelial cells. Lhx2 deficiency leads to baldness, because the niche can no longer anchor its hair and maintain the proper behavior of its SCs as a result of greatly perturbed HF-SCs polarization and niche architecture.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHX2
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. Q9Z0S2
Gene Symbols Lhx2
Host Species Rabbit
Immunogen Recombinant protein corresponding to mouse LHX2.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_034840
Target Species Human, Mouse
Content And Storage −20°C for one year
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.