Learn More
Description
Specifications
Specifications
| Antigen | LHX5 |
| Applications | Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Precipitated antibody in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives. |
| Gene Symbols | LHX5; MGC129689 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide, amino acids 148-164 from mouse LHX5. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.